Learn More
NIPSNAP1 Rabbit anti-Human, Mouse, Rat, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of NIPSNAP1.
Brand: Abnova PAB13354.100ug
Description
NIPSNAP1 protein has a strong sequence similarity to the central portion of a hypothetical protein encoded by C. elegans chromosome III between a 4-nitrophenylphosphatase (NIP) domain and non-neuronal SNAP25-like protein. The NIPSNAP1 gene is located between the NF2 and pK1.3 genes on 22q12. [provided by RefSeq
Specifications
| NIPSNAP1 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of NIPSNAP1. | |
| In PBS (0.02% sodium azide) | |
| NIPSNAP1 | |
| A synthetic peptide corresponding to internal region 19 amino acids of human NIPSNAP1. | |
| RUO | |
| 8508 | |
| Store at 4°C for three months. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| Immunohistochemistry (PFA fixed), Western Blot | |
| Unconjugated | |
| Western Blot (0.5-1 ug/mL) The optimal working dilution should be determined by the end user. | |
| NIPSNAP1 | |
| Rabbit | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only