Learn More
NLRP7, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody rasied against synthetic peptide of NLRP7.
Brand: Abnova PAB19305.100ug
Description
This gene encodes a member of the NACHT, leucine rich repeat, and PYD containing (NLRP) protein family. It has an N-terminal pyrin domain, followed by a NACHT domain, a NACHT-associated domain (NAD), and a C-terminal leucine-rich repeat (LRR) region. NLRP proteins are implicated in the activation of proinflammatory caspases through multiprotein complexes called inflammasomes. This gene may act as a feedback regulator of caspase-1-dependent interleukin 1-beta secretion. Alternative splicing results in multiple transcript variants encoding different isoforms. [provided by RefSeq
Specifications
| NLRP7 | |
| Polyclonal | |
| Unconjugated | |
| Western Blot (1 ug/mL) Immunohistochemistry (10 ug/mL) The optimal working dilution should be determined by the end user. | |
| NLRP7 | |
| NLRP7 | |
| A synthetic peptide corresponding to 18 amino acids near N-terminus of human NLRP7. | |
| 100 μg | |
| Primary | |
| Human | |
| Liquid |
| ELISA, Immunohistochemistry, Western Blot | |
| 1 mg/mL | |
| Rabbit polyclonal antibody rasied against synthetic peptide of NLRP7. | |
| In PBS (0.02% sodium azide) | |
| CLR19.4/FLJ94610/HYDM/MGC126470/MGC126471/NALP7/NOD12/PAN7/PYPAF3 | |
| Rabbit | |
| Peptide affinity purification | |
| RUO | |
| 199713 | |
| Store at 4°C for three months. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only