missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nos1, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of Nos1.
Brand: Abnova PAB26722.100ug
This item is not returnable.
View return policy
Description
Sequence: F-N-Sp-V-SSpecifications
| Nos1 | |
| Polyclonal | |
| Unconjugated | |
| Immunofluorescence (1:100-1:200) The optimal working dilution should be determined by the end user. | |
| Nos1 | |
| Nos1 | |
| A synthetic peptide (conjugated with KLH) corresponding to residues surrounding S852 of mouse Nos1. | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Liquid |
| Immunofluorescence | |
| 1 mg/mL | |
| Rabbit polyclonal antibody raised against synthetic peptide of Nos1. | |
| In PBS (without Mg2+ and Ca2+), 150mM NaCl, pH 7.4 (50% glycerol, 0.02% sodium azide) | |
| 2310005C01Rik/NO/NOS-I/Nos-1/bNOS/nNOS | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 18125 | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction