missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NR1H3 Rabbit anti-Human, Mouse, Rat, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of NR1H3.
Brand: Abnova PAB13685.100ug
This item is not returnable.
View return policy
Specifications
| NR1H3 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of NR1H3. | |
| Lyophilized from 0.9mg NaCl, 0.2mg Na2HPO4 (5mg BSA, 0.05mg sodium azide, 0.05mg Thimerosal) | |
| LXR-a/LXRA/RLD-1 | |
| Rabbit | |
| Affinity purification | |
| RUO | |
| 10062 | |
| Human, Mouse, Rat | |
| Lyophilized |
| Western Blot | |
| Unconjugated | |
| Western Blot (1 ug/mL) The optimal working dilution should be determined by the end user. | |
| NR1H3 | |
| NR1H3 | |
| A synthetic peptide corresponding to amino acids at C-terminus of human NR1H3. | |
| 100 μg | |
| Primary | |
| Identical to the related rat and mouse sequence. | |
| Store at -20°C on dry atmosphere. After reconstitution with 200μL of deionized water to a concentration of 500μg/mL store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction