missing translation for 'onlineSavingsMsg'
Learn More

Nr2c1, Rabbit, Polyclonal Antibody, Abnova™

Product Code. 16092487
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16092487 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16092487 Supplier Abnova Supplier No. PAB16944.100uL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit polyclonal antibody raised against partial recombinant Nr2c1.

Specifications

Antigen Nr2c1
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against partial recombinant Nr2c1.
Dilution Western Blot (1:1000) The optimal working dilution should be determined by the end user.
Formulation In serum
Gene Nr2c1
Gene Alias 4831444H07Rik/80.3/Eenr/TR2/Tr2-11
Gene Symbols Nr2c1
Host Species Rabbit
Immunogen Recombinant protein corresponding to N-terminus of mouse Nr2c1.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22025
Test Specificity This antibody has been directly tested for reactivity in mouse, human and rat. specific to the ∼64 KDa TR2 protein in Western blots of testes and nuclear extracts from MEL cell lines.
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.