Learn More
NR4A1 (phospho S351), Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic phosphopeptide of NR4A1.
Brand: Abnova PAB0547.400uL
Description
This gene encodes a member of the steroid-thyroid hormone-retinoid receptor superfamily. Expression is induced by phytohemagglutinin in human lymphocytes and by serum stimulation of arrested fibroblasts. The encoded protein acts as a nuclear transcription factor. Translocation of the protein from the nucleus to mitochondria induces apoptosis. Multiple alternatively spliced variants, encoding the same protein, have been identified. [provided by RefSeq
Specifications
| NR4A1 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic phosphopeptide of NR4A1. | |
| In PBS (0.09% sodium azide) | |
| GFRP1/HMR/MGC9485/N10/NAK-1/NGFIB/NP10/NUR77/TR3 | |
| Rabbit | |
| Protein A purification | |
| RUO | |
| 3164 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| Dot Blot, ELISA | |
| Unconjugated | |
| Dot Blot (1:100-500) The optimal working dilution should be determined by the end user. | |
| NR4A1 | |
| NR4A1 | |
| Synthetic phosphopeptide (conjugated with KLH) corresponding to residues surrounding S351 of human NR4A1. | |
| 400 μL | |
| Primary | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only