missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ntrk2, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of Ntrk2.
Brand: Abnova PAB0721.100uL
This item is not returnable.
View return policy
Description
Sequence: AFPRLEPNSIDPENCSpecifications
| Ntrk2 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of Ntrk2. | |
| Lyophilized from PBS | |
| RATTRKB1/TRKB1/Tkrb | |
| Rabbit | |
| 100 μL | |
| Primary | |
| Sequence was chosen to avoid cross reactivity with TrkA and TrkC. | |
| Store at 4°C on dry atmosphere. After reconstitution with deionized water store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
| Immunohistochemistry | |
| Unconjugated | |
| Immunohistochemistry (1:1000-1:3000) The optimal working dilution should be determined by the end user. | |
| Ntrk2 | |
| Ntrk2 | |
| A synthetic peptide (conjugated with KLH) corresponding to amino acids 23-36 of rat Ntrk2. | |
| RUO | |
| 25054 | |
| Human, Rat | |
| Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction