missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nup210, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against partial recombinant Nup210.
Brand: Abnova PAB12337.100uL
This item is not returnable.
View return policy
Specifications
| Nup210 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against partial recombinant Nup210. | |
| In antiserum (0.09% sodium azide) | |
| Pom210 | |
| Rabbit | |
| 100 μL | |
| Primary | |
| Rat Nup210 (C-terminal). | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| Immunofluorescence, Western Blot | |
| Unconjugated | |
| Western Blot (1:1000) Immunofluorescence (1:100) The optimal working dilution should be determined by the end user. | |
| Nup210 | |
| Nup210 | |
| Recombinant protein corresponding to C-terminus of rat Nup210. | |
| RUO | |
| 58958 | |
| Human, Monkey, Mouse, Rat | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction