missing translation for 'onlineSavingsMsg'
Learn More
Learn More
OPRD1 Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of OPRD1.
Brand: Abnova PAB25117.100uL
This item is not returnable.
View return policy
Specifications
| OPRD1 | |
| Polyclonal | |
| Unconjugated | |
| Western Blot (1:500-1:1000) The optimal working dilution should be determined by the end user. | |
| OPRD1 | |
| OPRD1 | |
| A synthetic peptide corresponding to OPRD1. | |
| 100 μL | |
| Primary | |
| OPRD1 polyclonal antibody detects endogenous levels of OPRD1 protein. | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| Western Blot | |
| 1 mg/mL | |
| Rabbit polyclonal antibody raised against synthetic peptide of OPRD1. | |
| In PBS, pH 7.2 (0.05% sodium azide) | |
| OPRD | |
| Rabbit | |
| Affinity purification | |
| RUO | |
| 4985 | |
| Human | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction