Learn More
PAK6 Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of PAK6.
Brand: Abnova PAB7884.100uL
Description
This gene encodes a member of the p21-activated kinase (PAK) family. The proteins of this family are Rac/Cdc42-associated Ste20-like Ser/Thr protein kinases, characterized by a highly conserved amino-terminal Cdc42/Rac interactive binding (CRIB) domain and a carboxyl-terminal kinase domain. PAK kinases are implicated in the regulation of a number of cellular processes, including cytoskeleton rearrangement, apoptosis and the MAP kinase signaling pathway. The protein encoded by this gene was found to interact with androgen receptor (AR), which is a steroid hormone-dependent transcription factor that is important for male sexual differentiation and development. This gene was found to be highly expressed in testis and prostate tissues and the encoded protein was shown to cotranslocate into the nucleus with AR in response to androgen. Alternatively spliced transcript variants encoding the same protein have been found for this gene. [provided by RefSeq
Specifications
| PAK6 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of PAK6. | |
| In PBS (50% glycerol, 1mg/mL BSA, 0.05% sodium azide) | |
| PAK5 | |
| Rabbit | |
| 100 μL | |
| Primary | |
| The sequence used has two amino acid differences compared to rat and mouse PAK6. | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| ELISA, Western Blot | |
| Unconjugated | |
| ELISA (1:2000) Western Blot (1:500) The optimal working dilution should be determined by the end user. | |
| PAK6 | |
| PAK6 | |
| A synthetic peptide corresponding to N-terminus of human PAK6. | |
| RUO | |
| 56924 | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only