Learn More
PANK1 Rabbit anti-Human, Mouse, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of PANK1.
Brand: Abnova PAB2717.400uL
Description
This gene encodes a protein belonging to the pantothenate kinase family. Pantothenate kinase is a key regulatory enzyme in the biosynthesis of coenzyme A (CoA) in bacteria and mammalian cells. It catalyzes the first committed step in the universal biosynthetic pathway leading to CoA and is itself subject to regulation through feedback inhibition by CoA. Alternative splicing has been observed at this locus and three variants, each encoding a distinct isoform, have been identified. [provided by RefSeq
Specifications
| PANK1 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of PANK1. | |
| In PBS (0.09% sodium azide) | |
| MGC24596/PANK/PANK1a/PANK1b | |
| Rabbit | |
| Protein G purification | |
| RUO | |
| 53354 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| ELISA, Western Blot | |
| Unconjugated | |
| Western Blot (1:1000) The optimal working dilution should be determined by the end user. | |
| PANK1 | |
| PANK1 | |
| A synthetic peptide (conjugated with KLH) corresponding to N-terminus of human PANK1. | |
| 400 μL | |
| Primary | |
| Human, Mouse | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only