Learn More
PARL, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of PARL.
Brand: Abnova PAB13061.100ug
Description
This gene encodes a mitochondrial integral membrane protein. Following proteolytic processing of this protein, a small peptide (P-beta) is formed and translocated to the nucleus. This gene may be involved in signal transduction via regulated intramembrane proteolysis of membrane-tethered precursor proteins. Variation in this gene has been associated with increased risk for type 2 diabetes. Alternative splicing results in multiple transcript variants encoding different isoforms. [provided by RefSeq
Specifications
| PARL | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of PARL. | |
| In PBS (0.02% sodium azide) | |
| PRO2207/PSARL/PSARL1/PSENIP2/RHBDS1 | |
| Rabbit | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Liquid |
| Immunocytochemistry, Western Blot | |
| Unconjugated | |
| Western Blot (1-2 ug/mL) The optimal working dilution should be determined by the end user. | |
| PARL | |
| PARL | |
| A synthetic peptide corresponding to N-terminus 15 amino acids of human PARL. | |
| RUO | |
| 55486 | |
| Store at 4°C for three months. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |