Learn More
PAWR Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of PAWR.
Brand: Abnova PAB14552.100uL
Description
The tumor suppressor WT1 represses and activates transcription. The protein encoded by this gene is a WT1-interacting protein that itself functions as a transcriptional repressor. It contains a putative leucine zipper domain which interacts with the zinc finger DNA binding domain of WT1. This protein is specifically upregulated during apoptosis of prostate cells. [provided by RefSeq
Sequence: MATGGYRTSSGLGGS, QENKTLLKVVGQLTRSpecifications
| PAWR | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of PAWR. | |
| In PBS (0.02% sodium azide) | |
| PAR4/Par-4 | |
| Rabbit | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
| Western Blot | |
| Unconjugated | |
| Western Blot (1:500-1:1000) The optimal working dilution should be determined by the end user. | |
| PAWR | |
| PAWR | |
| A mixture of synthetic peptide corresponding to amino acids at N-terminus and C-terminus of human PAWR. | |
| RUO | |
| 5074 | |
| Store at 4°C for short term. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only