Learn More
PCDH12, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of PCDH12.
Brand: Abnova PAB16714.100ug
Description
This gene belongs to the protocadherin gene family, a subfamily of the cadherin superfamily. The encoded protein consists of an extracellular domain containing 6 cadherin repeats, a transmembrane domain and a cytoplasmic tail that differs from those of the classical cadherins. The gene localizes to the region on chromosome 5 where the protocadherin gene clusters reside. The exon organization of this transcript is similar to that of the gene cluster transcripts, notably the first large exon, but no significant sequence homology exists. The function of this cellular adhesion protein is undetermined but mouse protocadherin 12 does not bind catenins and appears to have no affect on cell migration or growth. [provided by RefSeq
Specifications
| PCDH12 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of PCDH12. | |
| In PBS (0.02% sodium azide) | |
| VE-cadherin-2/VECAD2 | |
| Rabbit | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Liquid |
| ELISA, Immunohistochemistry, Western Blot | |
| Unconjugated | |
| Western Blot (1-2 ug/mL) The optimal working dilution should be determined by the end user. | |
| PCDH12 | |
| PCDH12 | |
| A synthetic peptide corresponding to N-terminus 17 amino acids of human PCDH12. | |
| RUO | |
| 51294 | |
| Store at 4°C for three months. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only