Learn More
PCSK6 Rabbit anti-Bovine, Horse, Human, Monkey, Rabbit, Rat, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of PCSK6.
Brand: Abnova PAB25583.50ug
Description
The protein encoded by this gene belongs to the subtilisin-like proprotein convertase family. The members of this family are proprotein convertases that process latent precursor proteins into their biologically active products. This encoded protein is a calcium-dependent serine endoprotease that can cleave precursor protein at their paired basic amino acid processing sites. Some of its substrates are - transforming growth factor beta related proteins, proalbumin, and von Willebrand factor. This gene is thought to play a role in tumor progression. Alternatively spliced transcript variants encoding different isoforms have been identified. [provided by RefSeq
Specifications
| PCSK6 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of PCSK6. | |
| In PBS (0.09% sodium azide) | |
| PACE4/SPC4 | |
| Rabbit | |
| Immunoaffinity chromatography | |
| RUO | |
| 5046 | |
| Bovine, Equine, Human, Monkey, Rabbit, Rat | |
| Liquid |
| ELISA, Immunohistochemistry (PFA fixed) | |
| Unconjugated | |
| Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (10 ug/mL) The optimal working dilution should be determined by the end user. | |
| PCSK6 | |
| PCSK6 | |
| A synthetic peptide corresponding to 14 amino acids at Cys-rich motif (CRM) region of human PCSK6. | |
| 50 μg | |
| Primary | |
| BLAST analysis of the peptide immunogen showed no homology with other human proteins. | |
| Store at 4°C. For long term storage store at -80°C. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only