missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PDE1A7, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of PDE1A7.
Brand: Abnova PAB24720.100ug
This item is not returnable.
View return policy
Description
Sequence: MIFRRLLDTEDELCSpecifications
| PDE1A7 | |
| Polyclonal | |
| Unconjugated | |
| WB (1:500-1:750) Immunostain (1:200) The optimal working dilution should be determined by the end user. | |
| PDE1A | |
| PDE1A | |
| A synthetic peptide corresponding to human PDE1A7. | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Liquid |
| Immunoprecipitation, Western Blot | |
| 0.57 mg/mL | |
| Rabbit polyclonal antibody raised against synthetic peptide of PDE1A7. | |
| In 50% antibody stabilization buffer contains Tris-glycine buffer, pH 7.4 (stabilizing proteins, cryogenic stabilizers, glycerol, 0.02% sodium azide) | |
| HCAM1/HSPDE1A/MGC26303 | |
| Rabbit | |
| Affinity Purification | |
| RUO | |
| 5136 | |
| Store at -20°C Aliquot to avoid repeated freezing and thawing. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction