Learn More
PGBD2 Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant PGBD2.
Brand: Abnova PAB22363.100uL
Description
The piggyBac family of proteins, found in diverse animals, are transposases related to the transposase of the canonical piggyBac transposon from the moth, Trichoplusia ni. This family also includes genes in several genomes, including human, that appear to have been derived from the piggyBac transposons. This gene belongs to the subfamily of piggyBac transposable element derived (PGBD) genes. The PGBD proteins appear to be novel, with no obvious relationship to other transposases, or other known protein families. The exact function of this gene is not known. Two transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq
Sequence: DYKVDESEEIIVCRWHDSSVVNICSNAVGIEPVRLTSRHSGAAKTRTQVHQPSLVKLYQEKVGGVGRMDQNIAKYKVKIRGMKWYSSSpecifications
| PGBD2 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against recombinant PGBD2. | |
| In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide) | |
| PGBD2 | |
| Recombinant protein corresponding to amino acids of human PGBD2. | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
| Immunohistochemistry (PFA fixed) | |
| Unconjugated | |
| Immunohistochemistry (1:10-1:20) The optimal working dilution should be determined by the end user. | |
| PGBD2 | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 267002 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
For Research Use Only