Learn More
PHB Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of PHB.
Brand: Abnova PAB2548.400uL
Description
Prohibitin is an evolutionarily conserved gene that is ubiquitously expressed. It is thought to be a negative regulator of cell proliferation and may be a tumor suppressor. Mutations in PHB have been linked to sporadic breast cancer. Prohibitin is expressed as two transcripts with varying lengths of 3' untranslated region. The longer transcript is present at higher levels in proliferating tissues and cells, suggesting that this longer 3' untranslated region may function as a trans-acting regulatory RNA. [provided by RefSeq]
Specifications
| PHB | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of PHB. | |
| In PBS (0.09% sodium azide) | |
| PHB1 | |
| Rabbit | |
| Protein A purification | |
| RUO | |
| 5245 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| ELISA, Western Blot | |
| Unconjugated | |
| Western Blot (1:1000) The optimal working dilution should be determined by the end user. | |
| PHB | |
| PHB | |
| A synthetic peptide (conjugated with KLH) corresponding to amino acids 189-219 at C-terminus of human PHB. | |
| 400 μL | |
| Primary | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only