missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phosphothreonine, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against Phosphothreonine.
Brand: Abnova PAB10352.100ug
This item is not returnable.
View return policy
Specifications
| Phosphothreonine | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against Phosphothreonine. | |
| In 20mM KH2PO4, 150mM NaCl, pH 7.2 | |
| Phosphothreonine conjugated with KLH. | |
| RUO | |
| Reactivity is specific to phosphothreonine and minimal cross reactivity is observed against phosphotyrosine or phosphoserine. | |
| Liquid |
| ELISA, Immunohistochemistry, Immunoprecipitation, Western Blot | |
| Unconjugated | |
| ELISA (0.5 ug/mL) Western Blot (2.0 ug/mL) Immunoprecipitation (10.0 ug/mg sample) The optimal working dilution should be determined by the end user. | |
| Rabbit | |
| 100 μg | |
| Primary | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?
For Research Use Only