missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phosphotyrosine Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against Phosphotyrosine.
Brand: Abnova PAB13767.100ug
This item is not returnable.
View return policy
Specifications
| Phosphotyrosine | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against Phosphotyrosine. | |
| In TBS (0.09% sodium azide) | |
| Phosphotyrosine conjugated with KLH. | |
| RUO | |
| Recognizes protein phosphorylated on tyrosine residues. Does not cross-react with phosphoserine or phosphothreonine. | |
| Liquid |
| ELISA, Immunoprecipitation, Western Blot | |
| Unconjugated | |
| ELISA (0.5 ug/mL) Western Blot (2 ug/mL) Immunoprecipitation (10 ug) The optimal working dilution should be determined by the end user. | |
| Rabbit | |
| 100 μg | |
| Primary | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction