Learn More
PI4K2A, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of PI4K2A.
Brand: Abnova PAB2131.400uL
Description
Phosphatidylinositolpolyphosphates (PtdInsPs) are centrally involved in many biologic processes, ranging from cell growth and organization of the actin cytoskeleton to endo- and exocytosis. PI4KII phosphorylates PtdIns at the D-4 position, an essential step in the biosynthesis of PtdInsPs (Barylko et al., 2001 [PubMed 11244087]).[supplied by OMIM
Specifications
| PI4K2A | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of PI4K2A. | |
| In PBS (0.09% sodium azide) | |
| DKFZp761G1923/PI4KII/PIK42A/RP11-548K23.6 | |
| Rabbit | |
| Protein G purification | |
| RUO | |
| 55361 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| ELISA, Immunofluorescence, Immunohistochemistry, Western Blot | |
| Unconjugated | |
| Western Blot (1:1000) Immunohistochemistry (1:50-100) Immunofluorescence (1:10-50) The optimal working dilution should be determined by the end user. | |
| PI4K2A | |
| PI4K2A | |
| A synthetic peptide (conjugated with KLH) corresponding to N-terminus of human PI4K2A. | |
| 400 μL | |
| Primary | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only