missing translation for 'onlineSavingsMsg'
Learn More

PIBF1, Rabbit, Polyclonal Antibody, Abnova™

Product Code. 16077427
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16077427 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16077427 Supplier Abnova Supplier No. PAB27239.100ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit polyclonal antibody raised against synthetic peptide of PIBF1.

Specifications

Antigen PIBF1
Applications ELISA, Immunofluorescence, Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of PIBF1.
Dilution Western Blot (1-2 ug/mL) Immunohistochemistry (5 ug/mL) Immunofluorescence (20 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In PBS (0.02% sodium azide)
Gene PIBF1
Gene Alias C13orf24/KIAA1008/PIBF/RP11-505F3.1
Gene Symbols PIBF1
Host Species Rabbit
Immunogen A synthetic peptide corresponding to 15 amino acids at C-terminus of human PIBF1.
Purification Method Peptide affinity purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10464
Test Specificity Multiple isoforms of PIBF1 exists as a result of alternative splicing event.
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C for three months. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.