missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PIK3R1/PIK3R2/PIK3R3, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of PIK3R1/PIK3R2/PIK3R3.
Brand: Abnova PAB26835.100ug
This item is not returnable.
View return policy
Description
Sequence: L-Yp- E-E-YSpecifications
| PIK3R1/PIK3R2/PIK3R3 | |
| Polyclonal | |
| Unconjugated | |
| Immunohistochemistry (1:50-1:100) The optimal working dilution should be determined by the end user. | |
| GRB1/p85/p85-ALPHA | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 5295/5296/8503 | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| Immunohistochemistry (PFA fixed) | |
| 1 mg/mL | |
| Rabbit polyclonal antibody raised against synthetic peptide of PIK3R1/PIK3R2/PIK3R3. | |
| In PBS, 150mM NaCl, pH 7.4 (50% glycerol, 0.02% sodium azide) | |
| PIK3R1 | |
| A synthetic peptide (conjugated with KLH) corresponding to residues surrounding Y467/Y199/Y464 of human PIK3R1/PIK3R2/PIK3R3. | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction