missing translation for 'onlineSavingsMsg'
Learn More

PIM2 Rabbit anti-Human, Polyclonal Antibody, Abnova™

Product Code. 16044407
Change view
Click to view available options
Quantity:
400 μL
Unit Size:
400µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16044407 400 μL 400µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16044407 Supplier Abnova Supplier No. PAB2305.400uL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit polyclonal antibody raised against synthetic peptide of PIM2.

PIM2 belongs to a family of protooncogenes that encode serine/threonine protein kinases (Mikkers et al., 2004 [PubMed 15199164]).[supplied by OMIM

Specifications

Antigen PIM2
Applications ELISA, Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of PIM2.
Dilution Western Blot (1:1000) Immunohistochemistry (1:50-100 Immunoprecipitation) The optimal working dilution should be determined by the end user.
Formulation In PBS (0.09% sodium azide)
Gene PIM2
Gene Symbols PIM2
Host Species Rabbit
Immunogen A synthetic peptide (conjugated with KLH) corresponding to C-terminus of human PIM2.
Purification Method Protein G purification
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11040
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.