Learn More
PIN1 Rabbit anti-Bovine, Dog, Human, Monkey, Mouse, Rat, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of PIN1.
Brand: Abnova PAB11244.100ug
Description
The human PIN1 gene encodes an essential nuclear peptidylprolyl cis-trans isomerase (PPIase; EC 5.2.1.8) involved in regulation of mitosis. PIN1 belongs to a class of PPIases that includes the E. coli parvulin, yeast Ess1, and Drosophila dodo (dod) gene products (Lu et al., 1996 [PubMed 8606777]).[supplied by OMIM
Specifications
| PIN1 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of PIN1. | |
| In 20mM KH2PO4, 150mM NaCl, pH 7.2 (0.01% sodium azide) | |
| DOD/UBL5 | |
| Rabbit | |
| 100 μg | |
| Primary | |
| Bovine, Canine, Human, Monkey, Mouse, Rat | |
| Liquid |
| ELISA, Immunoprecipitation, Western Blot | |
| Unconjugated | |
| ELISA (1:2500-1:10000) Western Blot (1:500-1:3000) The optimal working dilution should be determined by the end user. | |
| PIN1 | |
| PIN1 | |
| A synthetic peptide corresponding to internal region of human PIN1. | |
| RUO | |
| 5300 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only