Learn More
PIWIL1, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody rasied against synthetic peptide of PIWIL1.
Brand: Abnova PAB19332.100ug
Description
This gene encodes a member of the PIWI subfamily of Argonaute proteins, evolutionarily conserved proteins containing both PAZ and Piwi motifs that play important roles in stem cell self-renewal, RNA silencing, and translational regulation in diverse organisms. The encoded protein may play a role as an intrinsic regulator of the self-renewal capacity of germline and hematopoietic stem cells. [provided by RefSeq
Specifications
| PIWIL1 | |
| Polyclonal | |
| Unconjugated | |
| Western Blot (1 ug/mL) Immunohistochemistry (10 ug/mL) The optimal working dilution should be determined by the end user. | |
| PIWIL1 | |
| PIWIL1 | |
| A synthetic peptide corresponding to 18 amino acids near N-terminus of human PIWIL1. | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Liquid |
| ELISA, Immunohistochemistry, Western Blot | |
| 1 mg/mL | |
| Rabbit polyclonal antibody rasied against synthetic peptide of PIWIL1. | |
| In PBS (0.02% sodium azide) | |
| HIWI/MIWI/PIWI | |
| Rabbit | |
| Peptide affinity purification | |
| RUO | |
| 9271 | |
| Store at 4°C for three months. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only