Learn More
PKMYT1 (phospho S83), Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic phosphopeptide of human PKMYT1.
Brand: Abnova PAB29213.100uL
Description
The protein encoded by this gene is a member of the serine/threonine protein kinase family. This kinase preferentially phosphorylates and inactivates cell division cycle 2 protein (CDC2), and thus negatively regulates cell cycle G2/M transition. This kinase is associated with the membrane throughout the cell cycle. Its activity is highly regulated during the cell cycle. Protein kinases AKT1/PKB and PLK (Polo-like kinase) have been shown to phosphorylate and regulate the activity of this kinase. Alternatively spliced transcript variants encoding distinct isoforms have been reported. [provided by RefSeq
Specifications
| PKMYT1 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic phosphopeptide of human PKMYT1. | |
| In PBS, pH 7.4 (without Mg2+ and Ca2+), (50% glycerol, 0.02% sodium azide) | |
| DKFZp547K1610/FLJ20093/MYT1 | |
| Rabbit | |
| Affinity purification | |
| RUO | |
| 9088 | |
| Human | |
| Liquid |
| Immunohistochemistry (PFA fixed) | |
| Unconjugated | |
| Immunohistochemistry (1:50-1:100) The optimal working dilution should be determined by the end user. | |
| PKMYT1 | |
| PKMYT1 | |
| Synthetic phosphopeptide (conjugated with KLH) corresponding to residues surrounding S83 of human PKMYT1. | |
| 100 μL | |
| Primary | |
| This antibody detects endogenous levels of PKMYT1 only when phosphorylated at serine 83. | |
| Store at -20°C. Avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only