Learn More
PLA2G3 Rabbit anti-Bovine, Human, Monkey, Pig, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of PLA2G3.
Brand: Abnova PAB16527.50ug
Description
PLA2G3 belongs to the family of secreted phospholipase A2 (sPLA2; EC 3.1.1.4) proteins. These Ca(2+)-dependent lipolytic enzymes have a conserved Ca(2+)-binding loop and a his-asp dyad in the catalytic site (Murakami et al., 2003 [PubMed 12522102]).[supplied by OMIM
Specifications
| PLA2G3 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of PLA2G3. | |
| In PBS (0.09% sodium azide) | |
| GIII-SPLA2/SPLA2III | |
| Rabbit | |
| Immunoaffinity purification | |
| RUO | |
| 50487 | |
| Bovine, Human, Monkey, Pig | |
| Liquid |
| Immunohistochemistry (PFA fixed) | |
| Unconjugated | |
| Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (2.5 ug/mL) The optimal working dilution should be determined by the end user. | |
| PLA2G3 | |
| PLA2G3 | |
| A synthetic peptide (conjugated with KLH) corresponding to human PLA2G3. | |
| 50 μg | |
| Primary | |
| Internal domain of human. | |
| Store at 4°C. For long term storage store at -80°C. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only