missing translation for 'onlineSavingsMsg'
Learn More

PLA2R1 Rabbit anti-Human, Polyclonal Antibody, Abnova™

Product Code. 16166530
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16166530 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16166530 Supplier Abnova Supplier No. PAB28009.100uL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit polyclonal antibody raised against recombinant PLA2R1

Secretory phospholipases A2 (PLA2s) have been purified from a variety of mammalian tissues as well as from insect and snake venoms. The prototype group I PLA2, the pancreatic PLA2 (PLA2G1B; MIM 172410), is involved in digestion, smooth muscle contraction, and cell proliferation. The prototype group II PLA2, the inflammatory-type PLA2 (PLA2G2A; MIM 172411), is involved in inflammatory conditions and is upregulated by proinflammatory cytokines like tumor necrosis factor (TNF; MIM 191160) and interleukin-1 (e.g., IL1B; MIM 147720). Differential toxicity of snake venom Pla2 appears to be linked to a variety of high-affinity receptors in different organs (Ancian et al., 1995 [PubMed 7721806]).[supplied by OMIM

Sequence: EEKTWHEALRSCQADNSALIDITSLAEVEFLVTLLGDENASETWIGLSSNKIPVSFEWSNDSSVIFTNWHTLEPHIFPNRSQLCVSAEQSEGHWKVKNCEERLFYICKKAGHVLSDAESGCQEGWERHGGFCYKID

Specifications

Antigen PLA2R1
Applications Immunohistochemistry, Immunohistochemistry (Formalin/Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against recombinant PLA2R1
Dilution Immunohistochemistry (1:500-1:1000) The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.2, (40% glycerol, 0.02% sodium azide)
Gene PLA2R1
Gene Alias CLEC13C/PLA2-R/PLA2G1R/PLA2IR/PLA2R
Gene Symbols PLA2R1
Host Species Rabbit
Immunogen Recombinant protein corresponding to amino acids of human PLA2R1
Purification Method Antigen affinity purification
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22925
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.