Learn More
PLAU Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant PLAU.
Brand: Abnova PAB13618.100ug
Description
This gene encodes a serine protease involved in degradation of the extracellular matrix and possibly tumor cell migration and proliferation. A specific polymorphism in this gene may be associated with late-onset Alzheimer's disease and also with decreased affinity for fibrin-binding. This protein converts plasminogen to plasmin by specific cleavage of an Arg-Val bond in plasminogen. Plasmin in turn cleaves this protein at a Lys-Ile bond to form a two-chain derivative in which a single disulfide bond connects the amino-terminal A-chain to the catalytically active, carboxy-terminal B-chain. This two-chain derivative is also called HMW-uPA (high molecular weight uPA). HMW-uPA can be further processed into LMW-uPA (low molecular weight uPA) by cleavage of chain A into a short chain A (A1) and an amino-terminal fragment. LMW-uPA is proteolytically active but does not bind to the uPA receptor. Alternatively spliced transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq]
Specifications
| PLAU | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against recombinant PLAU. | |
| In 50mM sodium phosphate buffer, 150mM NaCl, pH 7.4 | |
| ATF/UPA/URK/UROKINASE/u-PA | |
| Rabbit | |
| 100 μg | |
| Primary | |
| This polyclonal antibody is specific to human uPA (urokinase) and is useful for fibrnolytic and cancer studies. It is provided in the following manner for added convenience: Serum, IgG Fraction, Affinity Purified as well as labeled with Biotin or FITC. This antibody reacts with both free and complexed forms of uPA. | |
| Store at -80°C. Aliquot to avoid repeated freezing and thawing. |
| ELISA, Western Blot | |
| Unconjugated | |
| The optimal working dilution should be determined by the end user. | |
| PLAU | |
| PLAU | |
| Recombinant protein corresponding to human PLAU. | |
| RUO | |
| 5328 | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only