Learn More
POFUT2 Rabbit anti-Human, Mouse, Polyclonal Antibody, Abnova™
Description
Fucose is typically found as a terminal modification of branched chain glycoconjugates, but it also exists in direct O-linkage to serine or threonine residues within cystine knot motifs in epidermal growth factor (EGF; MIM 131530)-like repeats or thrombospondin (THBS; see MIM 188060) type-1 repeats. POFUT2 is an O-fucosyltransferase that use THBS type-1 repeats as substrates (Luo et al., 2006 [PubMed 16464857]).[supplied by OMIM
Specifications
Specifications
| Antigen | POFUT2 |
| Applications | ELISA, Immunohistochemistry (PFA fixed) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Description | Rabbit polyclonal antibody raised against synthetic peptide of POFUT2. |
| Dilution | Immunohistochemistry (1:50-1:100) ELISA (1:40000) The optimal working dilution should be determined by the end user. |
| Formulation | In PBS, pH 7.4 (150mM NaCl, 0.02% sodium azide, 50% glycerol) |
| Gene | POFUT2 |
| Gene Alias | C21orf80/FUT13 |
| Gene Symbols | POFUT2 |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.