missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rabbit anti-Porcine IgG (H+L) Secondary Antibody, HRP, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB120-6777
This item is not returnable.
View return policy
Description
IgG (H+L) Polyclonal specifically detects IgG (H+L) in Porcine samples. It is validated for Western Blot, ELISA, Immunohistochemistry.
Specifications
| IgG (H+L) | |
| Polyclonal | |
| HRP | |
| Rabbit | |
| Multi-step | |
| RUO | |
| Secondary | |
| Pig | |
| Purified |
| Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| 2.0 mg/mL | |
| Western Blot 1:1000 - 1:5000, ELISA 1:50000 - 1:100000, Immunohistochemistry 1:500 - 1:2500 | |
| Rabbit anti-Porcine IgG (H+L) Secondary Antibody [HRP] was produced by repeated immunization with Swine IgG whole molecule in rabbit. | |
| 1 mg | |
| Adaptive Immunity, Immunology | |
| Assay by immunoelectrophoresis resulted in a single precipitin arc against anti-Peroxidase, anti-Rabbit Serum, Swine IgG and Swine Serum. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction