missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PPP1R13L, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of PPP1R13L.
Brand: Abnova PAB10243.100ug
This item is not returnable.
View return policy
Description
IASPP is one of the most evolutionarily conserved inhibitors of p53 (TP53; MIM 191170), whereas ASPP1 (MIM 606455) and ASPP2 (MIM 602143) are activators of p53.[supplied by OMIM
Specifications
| PPP1R13L | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of PPP1R13L. | |
| In 20mM KH2PO4, 150mM NaCl, pH 7.2 (0.01% sodium azide) | |
| IASPP/NKIP1/RAI | |
| Rabbit | |
| 100 μg | |
| Primary | |
| This affinity-purified antibody is directed against the human iASPP protein. Expect reactivity against both isoform 1 and isoform 2. | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| ELISA, Immunohistochemistry (PFA fixed), Western Blot | |
| Unconjugated | |
| ELISA (1:40000-1:160000) Western Blot (1:1000-1:5000) Immunohistochemistry (1:1000-1:5000) The optimal working dilution should be determined by the end user. | |
| PPP1R13L | |
| PPP1R13L | |
| A synthetic peptide corresponding to amino acids 780-797 of human PPP1R13L. | |
| RUO | |
| 10848 | |
| Bovine, Canine, Human, Mouse | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction