Learn More
PRDM1 Rabbit anti-Human, Mouse, Rat, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of PRDM1.
Brand: Abnova PAB13234.100ug
Description
This gene encodes a protein that acts as a repressor of beta-interferon gene expression. The protein binds specifically to the PRDI (positive regulatory domain I element) of the beta-IFN gene promoter. Transcription of this gene increases upon virus induction. Two alternatively spliced transcript variants that encode different isoforms have been reported. [provided by RefSeq
Specifications
| PRDM1 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of PRDM1. | |
| In PBS (0.02% sodium azide) | |
| BLIMP1/MGC118922/MGC118923/MGC118924/MGC118925/PRDI-BF1 | |
| Rabbit | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Liquid |
| Immunohistochemistry (PFA fixed), Western Blot | |
| Unconjugated | |
| Western Blot (0.5-1 ug/mL) The optimal working dilution should be determined by the end user. | |
| PRDM1 | |
| PRDM1 | |
| A synthetic peptide corresponding to N-terminus 17 amino acids of human PRDM1. | |
| RUO | |
| 639 | |
| Store at 4°C for three months. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only