Learn More
PRKD1 Rabbit anti-Human, Mouse, Rat, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of PRKD1.
Brand: Abnova PAB26697.100ug
Description
Members of the protein kinase C (PKC) family function in many extracellular receptor-mediated signal transduction pathways. See PRKCA (MIM 176960) for further background information. The PRKCM gene encodes a cytosolic serine-threonine kinase that binds to the trans-Golgi network and regulates the fission of transport carriers specifically destined to the cell surface.[supplied by OMIM
Sequence: E-K-Sp-F-RSpecifications
| PRKD1 | |
| Polyclonal | |
| Unconjugated | |
| Western Blot (1:500-1:1000) The optimal working dilution should be determined by the end user. | |
| PRKD1 | |
| PRKD1 | |
| A synthetic peptide corresponding to residues surrounding S738 of human PRKD1. | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| Rabbit polyclonal antibody raised against synthetic peptide of PRKD1. | |
| In PBS (without Mg2+ and Ca2+), 150mM NaCl, pH 7.4 (50% glycerol, 0.02% sodium azide) | |
| PKC-MU/PKCM/PKD/PRKCM | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 5587 | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only