missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Protein Phosphatase 2 C alpha/beta, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of Protein Phosphatase 2 C alpha/beta.
Brand: Abnova PAB10354.100ug
This item is not returnable.
View return policy
Specifications
| Protein Phosphatase 2 C alpha/beta | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of Protein Phosphatase 2 C alpha/beta. | |
| In PBS (0.08% sodium azide) | |
| A synthetic peptide (conjugated with KLH) corresponding to Protein Phosphatase 2 C alpha/beta. | |
| RUO | |
| Bovine, Human, Mouse, Rat | |
| Liquid |
| Immunoprecipitation, Western Blot | |
| Unconjugated | |
| Western blot (5 to 10 ug/mL) Immunoprecipitation (5 to 10 ug/mL) The optimal working dilution should be determined by the end user. | |
| Rabbit | |
| 100 μg | |
| Primary | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction