missing translation for 'onlineSavingsMsg'
Learn More

PRRT3, Rabbit, Polyclonal Antibody, Abnova™

Product Code. 16175380
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16175380 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16175380 Supplier Abnova Supplier No. PAB22717.100uL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit polyclonal antibody raised against recombinant PRRT3.

Sequence: PLENSEIPMIPGAHPKGSVGSEPQAFDVFPENPRADSHRNSDVRHAPAEEMPEKPVASPLGPALYGPKAAQGAQRERLPVTDDLQMAQGPSSH

Specifications

Antigen PRRT3
Applications Immunohistochemistry (PFA fixed)
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against recombinant PRRT3.
Dilution Immunohistochemistry (1:200-1:500) The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide)
Gene PRRT3
Gene Alias FLJ33674/MGC105134/MGC33990
Gene Symbols PRRT3
Host Species Rabbit
Immunogen Recombinant protein corresponding to amino acids of human PRRT3.
Purification Method Antigen affinity purification
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 285368
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.