Learn More
PTGES Rabbit anti-Human, Monkey, Mouse, Pig, Rabbit, Rat, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of PTGES.
Brand: Abnova PAB28173.50ug
Description
The protein encoded by this gene is a glutathione-dependent prostaglandin E synthase. The expression of this gene has been shown to be induced by proinflammatory cytokine interleukin 1 beta (IL1B). Its expression can also be induced by tumor suppressor protein TP53, and may be involved in TP53 induced apoptosis. Knockout studies in mice suggest that this gene may contribute to the pathogenesis of collagen-induced arthritis and mediate acute pain during inflammatory responses. [provided by RefSeq
Specifications
| PTGES | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of PTGES. | |
| In PBS (0.09% sodium azide) | |
| MGC10317/MGST-IV/MGST1-L1/MGST1L1/MPGES/PGES/PIG12/PP102/PP1294/TP53I12/mPGES-1 | |
| Rabbit | |
| Immunoaffinity chromatography | |
| RUO | |
| 9536 | |
| Human, Monkey, Mouse, Pig, Rabbit, Rat | |
| Liquid |
| Immunohistochemistry (PFA fixed) | |
| Unconjugated | |
| Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (5 ug/ml) | |
| PTGES | |
| PTGES | |
| A synthetic peptide corresponding to 16 amino acids at N-terminus region of human PTGES. | |
| 50 μg | |
| Primary | |
| BLAST analysis of the peptide immunogen showed no homology with other human proteins. | |
| Store at 4°C. For long term storage store at -80°C. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only