Learn More
PTH2R Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of PTH2R.
Brand: Abnova PAB26394.50ug
Description
The protein encoded by this gene is a member of the G-protein coupled receptor family 2. This protein is a receptor for parathyroid hormone (PTH). This receptor is more selective in ligand recognition and has a more specific tissue distribution compared to parathyroid hormone receptor 1 (PTHR1). It is activated only by PTH and not by parathyroid hormone-like hormone (PTHLH) and is particularly abundant in brain and pancreas. [provided by RefSeq
Specifications
| PTH2R | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of PTH2R. | |
| In PBS (0.09% sodium azide) | |
| PTHR2 | |
| Rabbit | |
| Immunoaffinity chromatography | |
| RUO | |
| 5746 | |
| Human | |
| Liquid |
| Immunohistochemistry (PFA fixed) | |
| Unconjugated | |
| Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (9 ug/mL) The optimal working dilution should be determined by the end user. | |
| PTH2R | |
| PTH2R | |
| A synthetic peptide corresponding to 20 amino acids at 1st extracellular domain of human PTH2R. | |
| 50 μg | |
| Primary | |
| BLAST analysis of the peptide immunogen showed no homology with other human proteins. | |
| Store at 4°C. For long term storage store at -80°C. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only