Learn More
PTPN9, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of PTPN9.
Brand: Abnova PAB4083.400uL
Description
The protein encoded by this gene is a member of the protein tyrosine phosphatase (PTP) family. PTPs are known to be signaling molecules that regulate a variety of cellular processes including cell growth, differentiation, mitotic cycle, and oncogenic transformation. This PTP contains an N-terminal domain that shares a significant similarity with yeast SEC14, which is a protein that has phosphatidylinositol transfer activity and is required for protein secretion through the Golgi complex in yeast. This PTP was found to be activated by polyphosphoinositide, and is thought to be involved in signaling events regulating phagocytosis. [provided by RefSeq
Specifications
| PTPN9 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of PTPN9. | |
| In PBS (0.09% sodium azide) | |
| MEG2/PTPMEG2 | |
| Rabbit | |
| Protein G purification | |
| RUO | |
| 5780 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| ELISA, Immunohistochemistry (PFA fixed), Western Blot | |
| Unconjugated | |
| ELISA (1:1000) Western Blot (1:100-500) Immunohistochemistry (1:10-50) The optimal working dilution should be determined by the end user. | |
| PTPN9 | |
| PTPN9 | |
| A synthetic peptide (conjugated with KLH) corresponding to C-terminus of human PTPN9. | |
| 400 μL | |
| Primary | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only