missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PXN Rabbit anti-Human, Mouse, Rat, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of PXN.
Brand: Abnova PAB7873.100uL
This item is not returnable.
View return policy
Specifications
| PXN | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of PXN. | |
| In PBS (50% glycerol, 1mg/mL BSA, 0.05% sodium azide) | |
| FLJ16691 | |
| Rabbit | |
| 100 μL | |
| Primary | |
| This peptide sequence is highly conserved in rat and mouse paxillins, and is found in all three paxillin isoforms (a, b, and g). | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| ELISA, Western Blot | |
| Unconjugated | |
| ELISA (1:2000) Western Blot (1:500) The optimal working dilution should be determined by the end user. | |
| PXN | |
| PXN | |
| A synthetic peptide (conjugated with KLH) corresponding to C-terminus of human PXN. | |
| RUO | |
| 5829 | |
| Human, Mouse, Rat | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction