Learn More
Raf1 (phospho S642), Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic phosphopeptide of Raf1.
Brand: Abnova PAB9660.100uL
This item is not returnable.
View return policy
Specifications
| Raf1 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic phosphopeptide of Raf1. | |
| In 10mM HEPES, 150mM NaCl, pH 7.5 (50% glycerol, 10% BSA) | |
| Raf1 | |
| Synthetic phosphopeptide corresponding to residues surrounding S642 of rat Raf1. | |
| 100 μL | |
| Primary | |
| Bovine, Canine, Chicken, Human, Mouse, Primate, Rat | |
| Liquid |
| Western Blot | |
| Unconjugated | |
| Western Blot (1:1000) The optimal working dilution should be determined by the end user. | |
| Raf1 | |
| Rabbit | |
| Affinity purification | |
| RUO | |
| 24703 | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |