Learn More
RANBP9, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against partial recombinant RANBP9.
Brand: Abnova PAB16671.100uL
Description
This gene encodes a protein that binds RAN, a small GTP binding protein belonging to the RAS superfamily that is essential for the translocation of RNA and proteins through the nuclear pore complex. The protein encoded by this gene has also been shown to interact with several other proteins, including met proto-oncogene, homeodomain interacting protein kinase 2, androgen receptor, and cyclin-dependent kinase 11. [provided by RefSeq
Specifications
| RANBP9 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against partial recombinant RANBP9. | |
| In antiserum (0.09% sodium azide) | |
| RANBPM | |
| Rabbit | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
| Immunocytochemistry, Immunoprecipitation, Western Blot | |
| Unconjugated | |
| Western Blot (1:2000) The optimal working dilution should be determined by the end user. | |
| RANBP9 | |
| RANBP9 | |
| Recombinant protein corresponding to the region from F133 to Y229 of human RANBP9. | |
| RUO | |
| 10048 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only