missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rabbit anti-Rat IgM Heavy Chain Secondary Antibody, Janelia Fluor 549, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-72790JF549
This item is not returnable.
View return policy
Description
IgM Heavy Chain Polyclonal antibody specifically detects IgM Heavy Chain in Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry
Specifications
| IgM Heavy Chain | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Multi-step | |
| RUO | |
| Secondary | |
| Rat | |
| IgG |
| Western Blot, ELISA, Immunohistochemistry | |
| Janelia Fluor 549 | |
| Constant region of heavy chain of IgM, Ig Mu Chain C Region BOT, Ig Mu Chain C Region GAL, Ig Mu Chain C Region OU, immunoglobulin heavy constant mu, Immunoglobulin mu chain | |
| Rat IgM whole molecule | |
| 0.5 mL | |
| B Cell Development and Differentiation Markers, Immunology | |
| 3507 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction