Learn More
RAX, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against partial recombinant RAX.
Brand: Abnova PAB0149.100ug
Description
Development of the vertebrate eye requires a series of steps including specification of the anterior neural plate, evagination of the optic vesicles from the ventral forebrain, and the cellular differentiation of the lens and retina. Homeobox-containing genes, including RAX, play a critical role in vertebrate and invertebrate eye formation (Mathers et al., 1997 [PubMed 9177348]).[supplied by OMIM
Specifications
| RAX | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against partial recombinant RAX. | |
| In PBS (0.05% BSA, 0.05% sodium azide) | |
| MCOP3/RX | |
| Rabbit | |
| 100 μg | |
| Primary | |
| Canine, Human, Monkey | |
| Liquid |
| Western Blot | |
| Unconjugated | |
| The optimal working dilution should be determined by the end user. | |
| RAX | |
| RAX | |
| Recombinant protein corresponding to amino acids 300-350 of human RAX. | |
| RUO | |
| 30062 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only