missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RHBDD2 Rabbit anti-Human, Mouse, Rat, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody rasied against synthetic peptide of RHBDD2.
Brand: Abnova PAB19162.100ug
This item is not returnable.
View return policy
Specifications
| RHBDD2 | |
| Polyclonal | |
| Unconjugated | |
| Western blot (1-2 ug/mL) The optimal working dilution should be determined by the end user. | |
| RHBDD2 | |
| NPD007/RHBDL7 | |
| Rabbit | |
| Affinity purification | |
| RUO | |
| 57414 | |
| Store at 4°C for three months. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| ELISA, Western Blot | |
| 1 mg/mL | |
| Rabbit polyclonal antibody rasied against synthetic peptide of RHBDD2. | |
| In PBS (0.02% sodium azide) | |
| NP_001035546 | |
| RHBDD2 | |
| A synthetic peptide corresponding to 18 amino acids near center region of human RHBDD2. | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction