missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rhov, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of Rhov.
Brand: Abnova PAB5927.100ug
This item is not returnable.
View return policy
Specifications
| Rhov | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of Rhov. | |
| In PBS, pH 7.2 (50% glycerol, 0.01% sodium azide) | |
| A030005A06Rik/Arhv/MGC29297 | |
| Rabbit | |
| 100 μg | |
| Primary | |
| Mouse, Rat | |
| Liquid |
| ELISA, Western Blot | |
| Unconjugated | |
| The optimal working dilution should be determined by the end user. | |
| Rhov | |
| Rhov | |
| A synthetic peptide corresponding to N-terminus of mouse Rhov. | |
| RUO | |
| 228543 | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
For Research Use Only
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur