missing translation for 'onlineSavingsMsg'
Learn More

RNF168, Rabbit, Polyclonal Antibody, Abnova™

Product Code. 16092337
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16092337 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16092337 Supplier Abnova Supplier No. PAB16741.100ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit polyclonal antibody raised against synthetic peptide of RNF168.

The complex repair response elicited by DNA double-strand breaks (DSBs) includes recruitment of several DNA repair proteins and ubiquitination of H2A-type histones (see MIM 142720). RNF168 is an E3 ubiquitin ligase critical for DSB repair (Stewart et al., 2009 [PubMed 19203578]).[supplied by OMIM

Specifications

Antigen RNF168
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of RNF168.
Dilution Western Blot (1-2 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In PBS (0.02% sodium azide)
Gene RNF168
Gene Alias FLJ35794
Gene Symbols RNF168
Host Species Rabbit
Immunogen A synthetic peptide corresponding to C-terminus 18 amino acids of human RNF168.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 165918
Target Species Human, Mouse
Content And Storage Store at 4°C for three months. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.