Learn More
RNF8, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody rasied against synthetic peptide of RNF8.
Brand: Abnova PAB19253.100ug
Description
The protein encoded by this gene contains a RING finger motif and a FHA domain. This protein has been shown to interact with several class II ubiquitin-conjugating enzymes (E2), including UBE2E1/UBCH6, UBE2E2, and UBE2E3, and may act as an ubiquitin ligase (E3) in the ubiquitination of certain nuclear proteins. Alternatively spliced transcript variants encoding distinct isoforms have been reported. [provided by RefSeq
Specifications
| RNF8 | |
| Polyclonal | |
| Unconjugated | |
| Western Blot (1-2 ug/mL) Immunofluorescence (20 ug/mL) The optimal working dilution should be determined by the end user. | |
| RNF8 | |
| RNF8 | |
| A synthetic peptide corresponding to 14 amino acids near C-terminus of human RNF8. | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Liquid |
| ELISA, Immunofluorescence, Western Blot | |
| 1 mg/mL | |
| Rabbit polyclonal antibody rasied against synthetic peptide of RNF8. | |
| In PBS (0.02% sodium azide) | |
| FLJ12013/KIAA0646 | |
| Rabbit | |
| Peptide affinity purification | |
| RUO | |
| 9025 | |
| Store at 4°C for three months. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only