missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ROBO4 Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of ROBO4.
Brand: Abnova PAB12048.100uL
This item is not returnable.
View return policy
Specifications
| ROBO4 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of ROBO4. | |
| In Tris-citrate/phosphate buffer, pH 7.0-8.0 (0.09% sodium azide) | |
| FLJ20798/MGC133352/MGC133353/MRB | |
| Rabbit | |
| 100 μL | |
| Primary | |
| This antibody recognizes the 85 KDa and 107 KDa isoforms of the ROBO4 human protein. | |
| Store at 4°C. Do not freeze. |
| Immunohistochemistry (PFA fixed), Western Blot | |
| Unconjugated | |
| Western Blot (1:500) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (5 ug/mL) The optimal working dilution should be determined by the end user. | |
| ROBO4 | |
| ROBO4 | |
| A synthetic peptide corresponding to amino acids 900-1007 of human ROBO4. | |
| RUO | |
| 54538 | |
| Human | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction